(S)-N-Boc-3-Amino-3-phenyl-1-propanol 97%

Supplier: Apollo Scientific

Synonyms: (S)-Boc-3-Amino-3-phenylpropan-1-ol

APOSOR471656-5GEA 159 EUR
APOSOR471656-5G
(S)-N-Boc-3-Amino-3-phenyl-1-propanol 97%
(S)-N-Boc-3-Amino-3-phenyl-1-propanol
Formula: C₁₄H₂₁NO₃
MW: 251,321 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025325
CAS Number: 718611-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR