Ethyl 5-amino-4-chloro-1-(4-methoxyphenyl)pyrazole-3-carboxylate

Supplier: Apollo Scientific

APOSOR480046-1GEA 797 EUR
APOSOR480046-1G
Ethyl 5-amino-4-chloro-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(4-methoxyphenyl)pyrazole-3-carboxylate
Formula: C₁₃H₁₄ClN₃O₃
MW: 295,722 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135217
CAS Number: 1427014-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR