Ethyl 5-amino-1-(3-sulphamoylphenyl)pyrazole-3-carboxylate

Supplier: Apollo Scientific

APOSOR480061-1GEA 1320 EUR
APOSOR480061-1G
Ethyl 5-amino-1-(3-sulphamoylphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(3-sulphamoylphenyl)pyrazole-3-carboxylate
Formula: C₁₂H₁₄N₄O₄S
MW: 310,329 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135152
CAS Number: 1427011-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR