Ethyl 5-amino-4-chloro-1-(4-pyridyl)pyrazole-3-carboxylate
Supplier: Apollo Scientific
APOSOR480069-1GEA
1200
EUR
APOSOR480069-1G
Ethyl 5-amino-4-chloro-1-(4-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(4-pyridyl)pyrazole-3-carboxylate
Formula:
C₁₁H₁₁ClN₄O₂ MW: 266,684 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23135162 CAS Number: 1427021-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...