1-(8-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile
Supplier: Apollo Scientific
APOSOR480152-1GEA
4540
EUR
APOSOR480152-1G
1-(8-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile
1-(8-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile
Formula:
C₁₄H₁₀BrN MW: 272,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30496595 CAS Number: 2114654-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...