5-Phenyl-3,4-dihydro-2H-pyrrole 94%
Supplier: Apollo Scientific
APOSOR480240-1GEA
524
EUR
APOSOR480240-1G
APOSOR480240-250MG
APOSOR480240-500MG
5-Phenyl-3,4-dihydro-2H-pyrrole 94%
5-Phenyl-3,4-dihydro-2H-pyrrole
Formula:
C₁₀H₁₁N MW: 145,201 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06661359 CAS Number: 700-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...