Amino-naphthalen-2-yl-acetic acid 95%
Supplier: Apollo Scientific
Synonyms:
2-Amino-2-(naphthalen-2-yl)acetic acid
APOSOR480472-1GEA
171
EUR
APOSOR480472-1G
Amino-naphthalen-2-yl-acetic acid 95%
Amino-naphthalen-2-yl-acetic acid
Formula:
C₁₂H₁₁NO₂ MW: 201,221 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02662462 CAS Number: 33741-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...