3-Chloro-4-cyanobenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
APOSOR480708-5GEA
960
EUR
APOSOR480708-5G
3-Chloro-4-cyanobenzenesulphonyl chloride 95%
3-Chloro-4-cyanobenzenesulphonyl chloride
Formula:
C₇H₃Cl₂NO₂S MW: 236,075 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09812743 CAS Number: 213130-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...