Ethyl 2-methyl-5-nitro-pyrazole-3-carboxylate 95%

Supplier: Apollo Scientific

APOSOR480849-1GEA 441 EUR
APOSOR480849-1G
Ethyl 2-methyl-5-nitro-pyrazole-3-carboxylate 95%
Ethyl 2-methyl-5-nitro-pyrazole-3-carboxylate
Formula: C₇H₉N₃O₄
MW: 199,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701484
CAS Number: 1245807-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR