8-Chloro-6-methoxyquinoline

Supplier: Apollo Scientific

APOSOR480860-1GEA 297 EUR
APOSOR480860-1G
8-Chloro-6-methoxyquinoline
8-Chloro-6-methoxyquinoline
Formula: C₁₀H₈ClNO
MW: 193,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907835
CAS Number: 796851-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR