2-(Chloromethyl)-4-hydroxyquinazoline

Supplier: Apollo Scientific
Warning

Synonyms: 2-(Chloromethyl)quinazlin-4-ol

APOSOR4920-1GEA 65.5 EUR
APOSOR4920-1G
2-(Chloromethyl)-4-hydroxyquinazoline
2-(Chloromethyl)-4-hydroxyquinazoline
Formula: C₉H₇ClN₂O
MW: 194,618 g/mol
Melting Pt: 249…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00665825
CAS Number: 3817-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR