3-Bromoprop-1-en-2-ylbenzene

Supplier: Apollo Scientific

APOSOR50994-100MGEA 495 EUR
APOSOR50994-100MG
3-Bromoprop-1-en-2-ylbenzene
3-Bromoprop-1-en-2-ylbenzene
Formula: C₉H₉Br
MW: 197,072 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09031921
CAS Number: 3360-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR