4,4'-Diphenyl-2,2'-bipyridine ≥97%
Supplier: Apollo Scientific
APOSOR510117-250MGEA
133
EUR
APOSOR510117-250MG
4,4'-Diphenyl-2,2'-bipyridine ≥97%
4,4'-Diphenyl-2,2'-bipyridine
Formula:
C₂₂H₁₆N₂ MW: 308,376 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00006421 CAS Number: 6153-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...