1-Chloro-6-methylisoquinoline-5-carboxylic acid ≥98%

Supplier: Apollo Scientific

APOSOR510158-100MGEA 483 EUR
APOSOR510158-100MG
1-Chloro-6-methylisoquinoline-5-carboxylic acid ≥98%
1-Chloro-6-methylisoquinoline-5-carboxylic acid
Formula: C₁₁H₈ClNO₂
MW: 221,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17014895
CAS Number: 1245643-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR