1-(7-Chloro-1H-indol-3-yl)-N,N-dimethylmethanamine ≥98%
Supplier: Apollo Scientific
APOSOR510200-100MGEA
754
EUR
APOSOR510200-100MG
1-(7-Chloro-1H-indol-3-yl)-N,N-dimethylmethanamine ≥98%
1-(7-Chloro-1H-indol-3-yl)-N,N-dimethylmethanamine
Formula:
C₁₁H₁₃ClN₂ MW: 208,687 g/mol Storage Temperature: Ambient |
CAS Number:
159503-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...