(S,S)-Me-BI-DIME ≥98%

Supplier: Apollo Scientific

APOSOR510255-100MGEA 297 EUR
APOSOR510255-100MG
(S,S)-Me-BI-DIME ≥98%
(S,S)-Me-BI-DIME
Formula: C₂₀H₂₅O₃P
MW: 344,384 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31707596
CAS Number: 1373432-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR