5-Bromo-6-methylpyridine-2-carboxylic acid 95+%

Supplier: Apollo Scientific

Synonyms: 5-Bromo-6-methylpicolinic acid3-Bromo-2-methylpyridine-6-carboxylic acid

APOSOR52563-1GEA 132 EUR
APOSOR52563-1G
5-Bromo-6-methylpyridine-2-carboxylic acid 95+%
5-Bromo-6-methylpyridine-2-carboxylic acid
Formula: C₇H₆BrNO₂
MW: 216,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181588
CAS Number: 137778-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR