2-Ethynyl-5-methoxypyrazine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Ethynyl-5-methoxy-1,4-diazine
, (5-Methoxypyrazin-2-yl)acetylene
APOSOR53017-250MGEA
165
EUR
APOSOR53017-250MG
2-Ethynyl-5-methoxypyrazine
2-Ethynyl-5-methoxypyrazine
Formula:
C₇H₆N₂O MW: 134,135 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22199320 CAS Number: 1374115-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...