Water, EMSURE® ACS, Reag. Ph. Eur. for analysis

1.04092.1000EA 151 EUR
1.04092.1000
Water, EMSURE® ACS, Reag. Ph. Eur. for analysis
Water
Formula: H₂O
MW: 18,02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR