3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt ≥98%

Supplier: Apollo Scientific
Warning

APOSBI5849-1GEA 11.3 EUR
APOSBI5849-1G APOSBI5849-5G
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt ≥98%
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt

Enzyme test reagent.


  • Assay (by titration) 98+%
  • Water (by Karl Fischer) < 3.0%

Formula: C₆H₃Cl₂NaO₄S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00009798
CAS Number: 54970-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR