Anisomycin (from Streptomyces griseolus)
Supplier: Apollo Scientific
|
Danger
|
APOSBIA4315-100MGEA
0
EUR
APOSBIA4315-100MG
APOSBIA4315-25MG
APOSBIA4315-250MG
Anisomycin (from Streptomyces griseolus)
Anisomycin
Formula:
C₁₄H₁₉NO₄ Storage Temperature: Freezer |
MDL Number:
MFCD00077650 CAS Number: 22862-76-6 EINECS: 245-269-7 UN: 3462 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...