2-(Benzyloxy)ethanol

Synonyms: Benzyl CellosolveEthylene Glycol Monobenzyl EtherBenzyl Glycol

APOSBIPG1184-2.5GEA 0 EUR
APOSBIPG1184-2.5G
2-(Benzyloxy)ethanol
2-(Benzyloxy)ethanol
Formula: C₉H₁₂O₂
MW: 152,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002868
CAS Number: 622-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR