Dimethyl sulphoxide-[D₆] (>99,8% D)
|
Warning
|
Synonyms:
DMSO-D6
, DMSO-d6
, Dimethyl sulphoxide-d6
, Dimethyl sulphoxide-[²H₆]
, Hexadeuterodimethyl sulphoxide
APOSDE60F-10X1MLEA
0
EUR
APOSDE60F-10X1ML
Dimethyl sulphoxide-[D₆] (>99,8% D)
Dimethyl sulphoxide-[D6]
Formula:
D₃CS(O)CD₃ MW: 84,09 g/mol Boiling Pt: 189 °C Melting Pt: 18,4 °C Density: 1,19 g/cm³ (25 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002090 CAS Number: 2206-27-1 EINECS: 218-617-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...