Dimethyl sulphoxide-[D₆] (>99,8% D) (max. 0.03% TMS (tetramethylsilan))

Supplier: Apollo Scientific

Synonyms: DMSO-D6DMSO-d6Dimethyl sulphoxide-d6Dimethyl sulphoxide-[²H₆]Hexadeuterodimethyl sulphoxide

APOSDE60JT-25MLEA 65.5 EUR
APOSDE60JT-25ML
Dimethyl sulphoxide-[D₆] (>99,8% D) (max. 0.03% TMS (tetramethylsilan))
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MW: 84,09 g/mol
Boiling Pt: 189 °C
Melting Pt: 18,4 °C
Density: 1,19 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR