Dimethyl sulphoxide-[D₆] (>99,5% D)

Supplier: Apollo Scientific
Warning

Synonyms: DMSO-D6DMSO-d6Dimethyl sulphoxide-d6Dimethyl sulphoxide-[²H₆]Hexadeuterodimethyl sulphoxide

APOSDE65B-50MLEA 77.1 EUR
APOSDE65B-50ML
Dimethyl sulphoxide-[D₆] (>99,5% D)
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MW: 84,09 g/mol
Boiling Pt: 189…190 °C
Melting Pt: 18…20 °C
Density: 1,19 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR