Dichloromethane-[D₂] (99.95% D)

Supplier: Apollo Scientific
Warning

Synonyms: Dichloromethane-[²H₂]Dideuteromethylene chlorideMethylene chloride-D2Methylene chloride-d2Dichloromethane-d2

APOSDE780-7.5MLEA 0 EUR
APOSDE780-7.5ML APOSDE780-10X0.75
Dichloromethane-[D₂] (99.95% D)
Dichloromethane-[D2]
Formula: CD₂Cl₂
MW: 86,92 g/mol
Boiling Pt: 40 °C
Melting Pt: –97 °C
Density: 1,362 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000882
CAS Number: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR