Rhodium(II) acetate dimer Rh 46,6%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Tetrakis(acetato)dirhodium(II)
, Dirhodium tetraacetate
APOSIN3003-250MGEA
285
EUR
APOSIN3003-250MG
Rhodium(II) acetate dimer Rh 46,6%
Rhodium (II) acetate dimer
Formula:
C₈H₁₂O₈Rh₂ MW: 441,987 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003538 CAS Number: 15956-28-2 EINECS: 240-084-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...