Ytterbium(III) sulphate octahydrate 99,9%

Supplier: Apollo Scientific

APOSIN3817-10GEA 68.9 EUR
APOSIN3817-10G
Ytterbium(III) sulphate octahydrate 99,9%
Ytterbium (III) sulphate octahydrate
Formula: O₁₂S₃Yb₂·8H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149887
CAS Number: 10034-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR