4-Bromo-3-methylbenzenesulphonamide 99%
Supplier: Apollo Scientific
|
Warning
|
APOSOR0115-250MGEA
14.7
EUR
APOSOR0115-250MG
4-Bromo-3-methylbenzenesulphonamide 99%
4-Bromo-3-methylbenzenesulphonamide
Formula:
C₇H₈BrNO₂S MW: 250,11 g/mol Melting Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD0394591 CAS Number: 879487-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...