4-Chloro-2-methylbenzonitrile

Supplier: Apollo Scientific
Warning

APOSOR0252-5GEA 15.1 EUR
APOSOR0252-5G
4-Chloro-2-methylbenzonitrile
4-Chloro-2-methylbenzonitrile
Formula: C₈H₆ClN
MW: 151,60
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00045603
CAS Number: 50712-68-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR