4-Morpholino-3-nitrobenzaldehyde

Supplier: Apollo Scientific
Warning

APOSOR0662-250MGEA 14.7 EUR
APOSOR0662-250MG
4-Morpholino-3-nitrobenzaldehyde
4-Morpholino-3-nitrobenzaldehyde
Formula: C₁₁H₁₂N₂O₄
Melting Pt: 53…55 °C
Storage Temperature: Ambient
CAS Number: 300541-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR