4-(Diethylcarbamoyl)phenylboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(N,N-Diethylaminocarbonyl)phenylboronic acid
, 4-(Diethylcarbamoyl)benzeneboronic acid
APOSOR10447-1GEA
14.7
EUR
APOSOR10447-1G
4-(Diethylcarbamoyl)phenylboronic acid 98%
4-(Diethylcarbamoyl)phenylboronic acid
Formula:
C₁₁H₁₆BNO₃ MW: 221,06 g/mol Melting Pt: 138…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411949 CAS Number: 389621-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...