4-(2-Aminoethyl)tetrahydropyran hydrochloride 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(Tetrahydro-2H-pyran-4-yl)ethanamine hydrochloride

APOSOR10492-1GEA 120 EUR
APOSOR10492-1G
4-(2-Aminoethyl)tetrahydropyran hydrochloride 98%
4-(2-Aminoethyl)tetrahydropyran hydrochloride
Formula: C₇H₁₆ClNO
MW: 165,66 g/mol
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD03411927
CAS Number: 389621-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR