4-Methyl-1,3-dioxane

APOSOR10967-100GEA 0 EUR
APOSOR10967-100G
4-Methyl-1,3-dioxane
4-Methyl-1,3-dioxane
Formula: C₅H₁₀O₂
MW: 102,1317 g/mol
Boiling Pt: 110…115 °C
Density: 0,98 g/cm³ (25 °C)
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00006569
CAS Number: 1120-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR