Ethyl-3-methyl-4-nitrobenzoate

Supplier: Apollo Scientific

APOSOR110362-5GEA 75.2 EUR
APOSOR110362-5G
Ethyl-3-methyl-4-nitrobenzoate
Ethyl-3-methyl-4-nitrobenzoate
Formula: C₁₀H₁₁NO₄
MW: 209,20
Storage Temperature: Ambient
MDL Number: MFCD00115804
CAS Number: 30650-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR