3',5'-Dichloro-2'-hydroxyacetophenone

APOSOR1118-25GEA 0 EUR
APOSOR1118-25G APOSOR1118-100G
3',5'-Dichloro-2'-hydroxyacetophenone
3',5'-Dichloro-2'-hydroxyacetophenone
Formula: C₈H₆Cl₂O₂
MW: 205,04 g/mol
Boiling Pt: 132…134 °C/18mmHg
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00016417
CAS Number: 3321-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR