4-Amino-6-hydroxypyrimidine

Supplier: Apollo Scientific

APOSOR11321-5GEA 82.9 EUR
APOSOR11321-5G
4-Amino-6-hydroxypyrimidine
4-Amino-6-hydroxypyrimidine
Formula: C₄H₅N₃O
MW: 111,10 g/mol
Melting Pt: 262…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00051502
CAS Number: 1193-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR