1,4-Dibromo-2,5-bis(bromomethyl)benzene 97%
|
Danger
|
APOSOR11479-5GEA
0
EUR
APOSOR11479-5G
APOSOR11479-1G
1,4-Dibromo-2,5-bis(bromomethyl)benzene 97%
1,4-Dibromo-2,5-bis(bromomethyl)benzene
Formula:
C₈H₆Br₄ MW: 421,7500 g/mol Melting Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411745 CAS Number: 35335-16-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...