5-Bromo-4-(4-methoxyphenyl)pyrimidine 98%

Supplier: Apollo Scientific

OR11511-1G OR11511-5G
APOSOR11511-1GEA 132 EUR
APOSOR11511-1G APOSOR11511-5G
5-Bromo-4-(4-methoxyphenyl)pyrimidine 98%
5-Bromo-4-(4-methoxyphenyl)pyrimidine
Formula: C₁₁H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09800946
CAS Number: 941294-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR