3-Chloro-(5-methoxycarbonyl)phenylboronic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: (3-Chloro-5-(methoxycarbonyl)phenyl)boronic acid3-Chloro-5-(methoxycarbonyl)phenylboronic acid

APOSOR11535-100MGEA 40.8 EUR
APOSOR11535-1G APOSOR11535-100MG APOSOR11535-250MG
3-Chloro-(5-methoxycarbonyl)phenylboronic acid 97%
3-Chloro-(5-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BClO₄
MW: 214,4107 g/mol
Melting Pt: 217…222 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09475906
CAS Number: 957120-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR