5-Bromo-4-hexylpyrimidine

APOSOR11582-1GEA 0 EUR
APOSOR11582-1G APOSOR11582-25G APOSOR11582-5G
5-Bromo-4-hexylpyrimidine
5-Bromo-4-hexylpyrimidine
Formula: C₁₀H₁₅BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09800936
CAS Number: 951884-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR