5-Bromo-4-isopentylpyrimidine

Supplier: Apollo Scientific

Synonyms: 5-Bromo-4-(3-methylbutyl)pyrimidine

OR11584-1G
APOSOR11584-1GEA 176 EUR
APOSOR11584-1G
5-Bromo-4-isopentylpyrimidine
5-Bromo-4-isopentylpyrimidine
Formula: C₉H₁₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09800938
CAS Number: 951884-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR