2-Bromo-N-isopropylbenzenesulphonamide
APOSOR11652-5GEA
0
EUR
APOSOR11652-5G
APOSOR11652-1G
2-Bromo-N-isopropylbenzenesulphonamide
2-Bromo-N-isopropylbenzenesulphonamide
Formula:
C₉H₁₂BrNO₂S Storage Temperature: Ambient |
MDL Number:
MFCD09800949 CAS Number: 951883-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...