6-Bromo-3-(ethylamino)[1,2,4]triazolo[4,3-a]pyridine
APOSOR11937-250MGEA
0
EUR
APOSOR11937-250MG
APOSOR11937-1G
6-Bromo-3-(ethylamino)[1,2,4]triazolo[4,3-a]pyridine
6-Bromo-3-(ethylamino)-[1,2,4]triazolo[4,3-a]pyridine
Formula:
C₈H₉BrN₄ Storage Temperature: Ambient |
MDL Number:
MFCD09801048 CAS Number: 951884-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...