2-Carboxy-4-chlorophenylboronic acid 98%
|
Warning
|
Synonyms:
2-Carboxy-4-chlorobenzeneboronic acid
APOSOR1202-1GEA
0
EUR
APOSOR1202-1G
2-Carboxy-4-chlorophenylboronic acid 98%
2-Carboxy-4-chlorophenylboronic acid
Formula:
C₇H₆BClO₄ MW: 200,39 g/mol Melting Pt: 300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659869 CAS Number: 850568-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...