Methyl-2-bromothiazole-4-carboxylate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 2-bromothiazole-4-carboxylate
APOSOR1218-1GEA
16.1
EUR
APOSOR1218-1G
Methyl-2-bromothiazole-4-carboxylate 97%
Methyl-2-bromothiazole-4-carboxylate
Formula:
C₅H₄BrNO₂S MW: 222,06 g/mol Melting Pt: 118…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659908 CAS Number: 170235-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...