6-Hydroxy-2-naphthaleneboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Hydroxynaphthalene-2-boronic acid
, (6-Hydroxynaphthalen-2-yl)boronic acid
APOSOR13171-1GEA
58.2
EUR
APOSOR13171-1G
6-Hydroxy-2-naphthaleneboronic acid 98%
6-Hydroxy-2-naphthaleneboronic acid
Formula:
C₁₀H₉BO₃ MW: 187,99 g/mol Melting Pt: 218…224 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00092986 CAS Number: 173194-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...