3,3-Dimethyl-2-oxobutyric acid 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Trimethylpyruvic Acid
, 3,3-Dimethyl-2-oxobutanoic acid
APOSOR13657-25GEA
136
EUR
APOSOR13657-25G
3,3-Dimethyl-2-oxobutyric acid 98%
3,3-Dimethyl-2-oxobutyric acid
Formula:
C₆H₁₀O₃ MW: 130,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00154352 CAS Number: 815-17-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...