6-Bromo-2-chlorobenzothiazole
Supplier: Apollo Scientific
|
Danger
|
APOSOR14084-5GEA
40.8
EUR
APOSOR14084-5G
6-Bromo-2-chlorobenzothiazole
6-Bromo-2-chlorobenzothiazole
Formula:
C₇H₃BrClNS MW: 248,53 g/mol Melting Pt: 98…103 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04971822 CAS Number: 80945-86-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...