4-(Isopropoxycarbonyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Isopropoxycarbonyl)benzeneboronic acid

APOSOR14156-5GEA 38.9 EUR
APOSOR14156-5G
4-(Isopropoxycarbonyl)phenylboronic acid 98%
4-(Isopropoxycarbonyl)phenylboronic acid
Formula: C₁₀H₁₃BO₄
MW: 208,02
Melting Pt: 111 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02259385
CAS Number: 342002-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR